missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
(S)-alpha-Methylphenylalanine hydrate, 98%, 98% ee, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC275431000
Specifications
(S)-α-Methylphenylalanine hydrate | |
Authentic | |
White | |
99%, (99+% e.e.) | |
97.5 | |
98.5% min. (HPLC) | |
2s-2-amino-2-methyl-3-phenylpropanoic acid, alpha-benzyl-l-ala, alpha-methyl-l-phe, alpha-methyl-l-phenylalanine, alpha-methylphenylalanine, l-alpha-methylphenylalanine, s-2-amimo-2-methyl-3-phenylpropanoic acid, s-2-amino-2-methyl-3-phenylpropanoic acid, s-2-amino-2-methyl-3-phenylpropionic acid, s-alpha-methylphenylalanine | |
CC(CC1=CC=CC=C1)(C(=O)O)N | |
179.22 | |
179.22 | |
98%,98% ee |
(HPLC)> 99.0% | |
− 18.00 | |
100mg | |
23239-35-2 | |
100.0 | |
C10H13NO2·xH2O | |
HYOWVAAEQCNGLE-JTQLQIEISA-N | |
(2S)-2-amino-2-methyl-3-phenylpropanoic acid | |
2724754 | |
Fine Crystalline Powder |
Chemical Identifiers
23239-35-2 | |
179.22 | |
2s-2-amino-2-methyl-3-phenylpropanoic acid, alpha-benzyl-l-ala, alpha-methyl-l-phe, alpha-methyl-l-phenylalanine, alpha-methylphenylalanine, l-alpha-methylphenylalanine, s-2-amimo-2-methyl-3-phenylpropanoic acid, s-2-amino-2-methyl-3-phenylpropanoic acid, s-2-amino-2-methyl-3-phenylpropionic acid, s-alpha-methylphenylalanine | |
(2S)-2-amino-2-methyl-3-phenylpropanoic acid |
C10H13NO2·xH2O | |
HYOWVAAEQCNGLE-JTQLQIEISA-N | |
2724754 | |
CC(CC1=CC=CC=C1)(C(=O)O)N |
Safety and Handling
TSCA : TSCA