missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
S-Methylisothiouronium sulfate, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1104422
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
S-Methylisothiouronium sulfate | |
∼235°C (decomposition) | |
C4H14N4O4S3 | |
100 g | |
Hygroscopic | |
BZZXQZOBAUXLHZ-UHFFFAOYSA-N | |
methyl carbamimidothioate;sulfuric acid | |
13347 | |
≥98% |
867-44-7 | |
1.28 | |
MFCD00013055 | |
3917217 | |
s-methylisothiourea hemisulfate, 2-methyl-2-thiopseudourea hemisulfate, carbamimidothioic acid, methyl ester, sulfate 2:1, s-methylisothiourea sulfate 2:1, s-methylthiouronium sulfate 2:1, bis 2-methylisothiouronium sulphate, methyl carbamimidothioate sulfate 2:1, methylcarbamimidothioate sulfate 2:1, s-methylisothiourea, sulfate, 2-methyl-2-thiopseudourea sulfate 2:1 | |
CSC(=N)N.CSC(=N)N.OS(=O)(=O)O | |
278.36 | |
278.38 |
Chemical Identifiers
867-44-7 | |
278.36 | |
BZZXQZOBAUXLHZ-UHFFFAOYSA-N | |
13347 | |
CSC(=N)N.CSC(=N)N.OS(=O)(=O)O |
C4H14N4O4S3 | |
MFCD00013055 | |
s-methylisothiourea hemisulfate, 2-methyl-2-thiopseudourea hemisulfate, carbamimidothioic acid, methyl ester, sulfate 2:1, s-methylisothiourea sulfate 2:1, s-methylthiouronium sulfate 2:1, bis 2-methylisothiouronium sulphate, methyl carbamimidothioate sulfate 2:1, methylcarbamimidothioate sulfate 2:1, s-methylisothiourea, sulfate, 2-methyl-2-thiopseudourea sulfate 2:1 | |
methyl carbamimidothioate;sulfuric acid |
Safety and Handling
GHS H Statement
H301
Toxic if swallowed.
P264b-P270-P301+P312-P330-P501c
H302
EINECSNumber : 212-759-7
RTECSNumber : UM9200000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only