Please login to your online account to display your discounted pricing

(S)-Methylisothiourea sulfate, Tocris


Highly selective iNOS inhibitor

Quantity 10mg

Manufacturer: r & d systems  RDS077610

 View more versions of this product

Catalog No. 077610

  • / Each
 

Description & Specifications

Chemical Identifiers

Chemical Name or Material (S)-Methylisothiourea sulfate
CAS 867-44-7
Recommended Storage Store at Room Temperature
Assay Percent Range >98%
Molecular Formula C4H14N4O4S3
Formula Weight 278.36
Quantity 10mg

Specifications

Chemical Name or Material (S)-Methylisothiourea sulfate
Assay Percent Range >98%
Molecular Formula C4H14N4O4S3
Formula Weight 278.36
Accessories

Store at Room Temperature