Please login to your online account to display your discounted pricing

(S)-Methylisothiourea sulfate, Tocris

Highly selective iNOS inhibitor

$113.57 - $478.51

Chemical Identifiers

Chemical Name or Material (S)-Methylisothiourea sulfate
Assay Percent Range >98%
Molecular Formula C4H14N4O4S3
Formula Weight 278.36
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

077610

 
r & d systems
RDS077610
10mg Each for $113.57

077650

 
r & d systems
RDS077650
50mg Each for $478.51
Description & Specifications

Specifications

Quantity 10mg
Formula Weight 278.36
Chemical Name or Material (S)-Methylisothiourea sulfate
Recommended Storage Store at Room Temperature
Molecular Formula C4H14N4O4S3
CAS 867-44-7
Assay Percent Range >98%
Safety and Handling

Store at Room Temperature