missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

S-Methylisothiourea Sulfate, TCI America™

Manufacturer:  TCI America M0442500G

 View more versions of this product

Catalog No. M0442500G



Specifications

Specifications

S-Methylisothiourea Sulfate
500g
C4H14N4O4S3
s-methylisothiourea hemisulfate, 2-methyl-2-thiopseudourea hemisulfate, carbamimidothioic acid, methyl ester, sulfate 2:1, s-methylisothiourea sulfate 2:1, s-methylthiouronium sulfate 2:1, bis 2-methylisothiouronium sulphate, methyl carbamimidothioate sulfate 2:1, methylcarbamimidothioate sulfate 2:1, s-methylisothiourea, sulfate, 2-methyl-2-thiopseudourea sulfate 2:1
CSC(=N)N.CSC(=N)N.OS(=O)(=O)O
278.36
278.36
≥98.0% (T)
White
867-44-7
MFCD00013055
BZZXQZOBAUXLHZ-UHFFFAOYSA-N
methyl carbamimidothioate;sulfuric acid
13347
Crystal-Powder at 20°C

Chemical Identifiers

867-44-7
278.36
BZZXQZOBAUXLHZ-UHFFFAOYSA-N
13347
CSC(=N)N.CSC(=N)N.OS(=O)(=O)O
C4H14N4O4S3
MFCD00013055
s-methylisothiourea hemisulfate, 2-methyl-2-thiopseudourea hemisulfate, carbamimidothioic acid, methyl ester, sulfate 2:1, s-methylisothiourea sulfate 2:1, s-methylthiouronium sulfate 2:1, bis 2-methylisothiouronium sulphate, methyl carbamimidothioate sulfate 2:1, methylcarbamimidothioate sulfate 2:1, s-methylisothiourea, sulfate, 2-methyl-2-thiopseudourea sulfate 2:1
methyl carbamimidothioate;sulfuric acid
Safety and Handling

Safety and Handling

EINECSNumber : (2)-2865&(9)-1413

RTECSNumber : UM9200000

TSCA : Yes

Documents