missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(S)-alpha-Methylaspartic acid, 98%, 98% ee, Thermo Scientific™

Chemical Identifiers

CAS 3227-17-6
Molecular Formula C5H8NO4
Molecular Weight (g/mol) 146.12
MDL Number MFCD00145254
InChI Key CWAYDJFPMMUKOI-YFKPBYRVSA-M
Synonym 2-methyl-l-aspartic acid, 2s-2-amino-2-methylbutanedioic acid, a-methyl-l-asp, alpha-methyl aspartic acid, alpha-methyl-l-asp, alpha-methylaspartic acid, ambotzhaa7550, aspartic acid, 2-methyl, h-a-me-asp-oh, s-+-2-amino-2-methylbutanedioic acid
PubChem CID 2724884
IUPAC Name (2S)-2-amino-2-methylbutanedioic acid
SMILES C[C@]([NH3+])(CC([O-])=O)C([O-])=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC290135000
SDS View Documents Promotion Details
Thermo Fisher Scientific
290135000
500mg N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

3227-17-6
146.12
CWAYDJFPMMUKOI-YFKPBYRVSA-M
2724884
C[C@]([NH3+])(CC([O-])=O)C([O-])=O
C5H8NO4
MFCD00145254
2-methyl-l-aspartic acid, 2s-2-amino-2-methylbutanedioic acid, a-methyl-l-asp, alpha-methyl aspartic acid, alpha-methyl-l-asp, alpha-methylaspartic acid, ambotzhaa7550, aspartic acid, 2-methyl, h-a-me-asp-oh, s-+-2-amino-2-methylbutanedioic acid
(2S)-2-amino-2-methylbutanedioic acid

Specifications

3227-17-6
100.0
>245.0°C
97.5% min
Authentic
MFCD00145254
500mg
CWAYDJFPMMUKOI-YFKPBYRVSA-M
+ 51.00
146.12
147.13
98%,98% ee
97.5
White
98.0% min. (HPLC)
C5H8NO4
HOOCC(CH3)NH2CH2COOH
Glass bottle
2-methyl-l-aspartic acid, 2s-2-amino-2-methylbutanedioic acid, a-methyl-l-asp, alpha-methyl aspartic acid, alpha-methyl-l-asp, alpha-methylaspartic acid, ambotzhaa7550, aspartic acid, 2-methyl, h-a-me-asp-oh, s-+-2-amino-2-methylbutanedioic acid
C[C@]([NH3+])(CC([O-])=O)C([O-])=O
(2S)-2-amino-2-methylbutanedioic acid
2724884
Fine Crystalline Powder or Powder
(S)-α-Methylaspartic acid, 99%
Safety and Handling

Safety and Handling

TSCA : TSCA

SDS
Product Certifications

RUO – Research Use Only