Please login to your online account to display your discounted pricing

Alfa Aesar™ S-Methyl pyridine-4-carbothioimidate hydriodide, 96%

Chemical Identifiers

Chemical Name or Material S-Methyl pyridine-4-carbothioimidate hydriodide
Molecular Formula C7H8N2S·HI
Formula Weight 280.13
MDL Number MFCD12026517
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5177403

  SDS
alfa aesar™
H5177403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 280.13
Chemical Name or Material S-Methyl pyridine-4-carbothioimidate hydriodide
Assay 96%
Molecular Formula C7H8N2S·HI
MDL Number MFCD12026517
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Melting Point 182° to 186°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.