Please login to your online account to display your discounted pricing

S-Methyl-L-cysteine, 99+%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material S-Methyl-L-cysteine
Assay 99.0% min.
Assay Percent Range 99+%
Molecular Formula C4H9NO2S
Linear Formula CH3SCH2CH(NH2)CO2H
Formula Weight 135.18g/mol
MDL Number MFCD00002612
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC226920100

  SDS
fisher scientific acros
226920100
Glass bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Infrared Spectrum Authentic
Packaging Glass bottle
Water 0.5% max. (K.F.)
Specific Rotation -28.5° to -30.5° (20°C, 589nm) (c=1, H2O)
Specific Rotation Condition − 29.50 (20.00°C c=1,water)
Color White to light beige
Quantity 10g
Chemical Name or Material S-Methyl-L-cysteine
CAS 1187-84-4
Physical Form White to light beige fine crystalline powder
Assay 99.0% min.
Assay Percent Range 99+%
Molecular Formula C4H9NO2S
Linear Formula CH3SCH2CH(NH2)CO2H
Formula Weight 135.18g/mol
MDL Number MFCD00002612
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.