Please login to your online account to display your discounted pricing

S-Methyl-L-cysteine, 99+%, ACROS Organics™


Quantity 10g
Packaging Glass bottle

Manufacturer: fisher scientific acros  226920100

 View more versions of this product

Catalog No. AC226920100

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Infrared Spectrum Authentic
Packaging Glass bottle
Water 0.5% max. (K.F.)
Specific Rotation -28.5° to -30.5° (20°C, 589nm) (c=1, H2O)
Specific Rotation Condition − 29.50 (20.00°C c=1,water)
Color White to light beige
Quantity 10g
Chemical Name or Material S-Methyl-L-cysteine
CAS 1187-84-4
Physical Form White to light beige fine crystalline powder
Assay 99.0% min.
Assay Percent Range 99+%
Molecular Formula C4H9NO2S
Linear Formula CH3SCH2CH(NH2)CO2H
Formula Weight 135.18g/mol
MDL Number MFCD00002612
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material S-Methyl-L-cysteine
Assay 99.0% min.
Assay Percent Range 99+%
Molecular Formula C4H9NO2S
Linear Formula CH3SCH2CH(NH2)CO2H
Formula Weight 135.18g/mol
MDL Number MFCD00002612
Accessories

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.

SDS & Safety Documents