missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
(S)-(+)-α-Methoxy-α-(trifluoromethyl)phenylacetyl chloride, 99%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC264112500
Description
Chiral derivatization reagentSpecifications
(S)-(+)-α-Methoxy-α-(trifluoromethyl)phenylacetyl chloride | |
89°C | |
Glass bottle | |
1.34 | |
+ 132.00 | |
Colorless to Yellow | |
20445-33-4 | |
100.0 | |
C10H8ClF3O2 | |
15, 6365 | |
PAORVUMOXXAMPL-SECBINFHSA-N | |
(2S)-3,3,3-trifluoro-2-methoxy-2-phenylpropanoyl chloride | |
2724611 | |
Liquid |
1.3400g/mL | |
Authentic | |
1.4680 to 1.4700 | |
+132° (20°C c=6,CHCl3) | |
213.0°C to 214.0°C | |
250mg | |
98.5 | |
98.5% min. (GC) | |
MFCD00067105 | |
+-mosher's chloride, +-mtpa chloride, +-mtpa-cl, 2s-3,3,3-trifluoro-2-methoxy-2-phenylpropanoyl chloride, s-+-a-methoxy-a-trifluoromethylphenylacetyl chloride, s-+-alpha-methoxy-alpha-trifluoromethyl phenylacetyl chloride, s-+-alpha-methoxy-alpha-trifluoromethylphenylacetyl chloride, s-+-alpha-methoxy-alpha-trifluoromethylphenylacetylchloride, s-3,3,3-trifluoro-2-methoxy-2-phenylpropanoyl chloride, unii-472va70ul4 | |
COC(C1=CC=CC=C1)(C(=O)Cl)C(F)(F)F | |
252.62 | |
252.62 | |
99% |
Chemical Identifiers
20445-33-4 | |
252.62 | |
PAORVUMOXXAMPL-SECBINFHSA-N | |
2724611 | |
COC(C1=CC=CC=C1)(C(=O)Cl)C(F)(F)F |
C10H8ClF3O2 | |
MFCD00067105 | |
+-mosher's chloride, +-mtpa chloride, +-mtpa-cl, 2s-3,3,3-trifluoro-2-methoxy-2-phenylpropanoyl chloride, s-+-a-methoxy-a-trifluoromethylphenylacetyl chloride, s-+-alpha-methoxy-alpha-trifluoromethyl phenylacetyl chloride, s-+-alpha-methoxy-alpha-trifluoromethylphenylacetyl chloride, s-+-alpha-methoxy-alpha-trifluoromethylphenylacetylchloride, s-3,3,3-trifluoro-2-methoxy-2-phenylpropanoyl chloride, unii-472va70ul4 | |
(2S)-3,3,3-trifluoro-2-methoxy-2-phenylpropanoyl chloride |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
GHS Signal Word: Danger