Please login to your online account to display your discounted pricing

(S)-MCPG, Tocris

Non-selective mGlu antagonist

$330.06 - $1,362.21

Chemical Identifiers

Chemical Name or Material (S)-MCPG
Assay Percent Range >99%
Molecular Formula C10H11NO4
Formula Weight 209.2
Synonym (S)-α-Methyl-4-carboxyphenylglycine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

033710

 
r & d systems
RDS033710
10mg Each for $330.06

033750

 
r & d systems
RDS033750
50mg Each for $1,362.21
Description & Specifications

Specifications

Quantity 10mg
Formula Weight 209.2
Chemical Name or Material (S)-MCPG
Assay Percent Range >99%
CAS 150145-89-4
Molecular Formula C10H11NO4
Synonym (S)-α-Methyl-4-carboxyphenylglycine
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature