missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(S)-(+)-Mandelic Acid ≥99%, Thermo Scientific Chemicals

$44.19 - $150.66

Chemical Identifiers

CAS 17199-29-0
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.149
MDL Number MFCD00004495
InChI Key IWYDHOAUDWTVEP-ZETCQYMHSA-N
Synonym s-+-mandelic acid, s-mandelic acid, s-2-hydroxy-2-phenylacetic acid, l-mandelic acid, l-+-mandelic acid, 2s-2-hydroxy-2-phenylacetic acid, mandelic acid, s, unii-l0umw58g3t, l +-mandelic acid
PubChem CID 439616
ChEBI CHEBI:32800
IUPAC Name (2S)-2-hydroxy-2-phenylacetic acid
SMILES C1=CC=C(C=C1)C(C(=O)O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC125300250
SDS View Documents Promotion Details
Thermo Scientific Chemicals
125300250
25 g Plastic bottle
Each for $44.19
 
AC125301000
SDS View Documents Promotion Details
Thermo Scientific Chemicals
125301000
100 g Plastic bottle
Each for $150.66
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

17199-29-0
152.149
IWYDHOAUDWTVEP-ZETCQYMHSA-N
439616
(2S)-2-hydroxy-2-phenylacetic acid
C8H8O3
MFCD00004495
s-+-mandelic acid, s-mandelic acid, s-2-hydroxy-2-phenylacetic acid, l-mandelic acid, l-+-mandelic acid, 2s-2-hydroxy-2-phenylacetic acid, mandelic acid, s, unii-l0umw58g3t, l +-mandelic acid
CHEBI:32800
C1=CC=C(C=C1)C(C(=O)O)O

Specifications

130°C to 135°C
99.0
Beige-Brown to White
C8H8O3
MFCD00004495
s-+-mandelic acid, s-mandelic acid, s-2-hydroxy-2-phenylacetic acid, l-mandelic acid, l-+-mandelic acid, 2s-2-hydroxy-2-phenylacetic acid, mandelic acid, s, unii-l0umw58g3t, l +-mandelic acid
C1=CC=C(C=C1)C(C(=O)O)O
152.149
CHEBI:32800
≥99.0% (Acid)
Authentic
Crystalline Powder
17199-29-0
100.0
99+%
C6H5CH(OH)CO2H
10, 192
IWYDHOAUDWTVEP-ZETCQYMHSA-N
(2S)-2-hydroxy-2-phenylacetic acid
439616
152.15
25 g
Plastic bottle
(S)-(+)-Mandelic acid, 99+%
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

LIGHT SENSITIVE

EINECSNumber : 241-240-8

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only