Please login to your online account to display your discounted pricing

S-Isopropylisothiourea hydrobromide, Tocris


iNOS inhibitor, acts at arginine binding site

Quantity 10mg

Manufacturer: r & d systems  RDS089710

 View more versions of this product

Catalog No. 089710

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 10mg
Formula Weight 199.11
Chemical Name or Material S-Isopropylisothiourea hydrobromide
Assay Percent Range >98%
Molecular Formula C4H10N2S·HBr
CAS 4269-97-0
Recommended Storage Store at Room Temperature

Specifications

Chemical Name or Material S-Isopropylisothiourea hydrobromide
Assay Percent Range >98%
Molecular Formula C4H10N2S·HBr
Formula Weight 199.11
Accessories

Store at Room Temperature