Learn More
(S)-(-)-Indoline-2-carboxylic acid, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 342080050
Specifications
(S)-(-)-Indoline-2-carboxylic acid | |
79815-20-6 | |
100.0 | |
98% | |
C9H9NO2 | |
5g | |
-112.5 | |
QNRXNRGSOJZINA-QMMMGPOBSA-N | |
(2S)-2,3-dihydro-1H-indole-2-carboxylic acid | |
2733920 | |
98% |
98% | |
97.5 | |
Authentic | |
Glass bottle | |
MFCD00070578 | |
−112.5° (20°C c=1,1N HCl) | |
s---indoline-2-carboxylic acid, s-indoline-2-carboxylic acid, 2s-2,3-dihydro-1h-indole-2-carboxylic acid, h-idc-oh, l-indoline-2-carboxylic acid, l---indoline-2-carboxylic acid, s-2,3-dihydro-1h-indole-2-carboxylic acid, s---indolin-2-carboxylic acid, 1h-indole-2-carboxylic acid, 2,3-dihydro-, 2s | |
C1C(NC2=CC=CC=C21)C(=O)O | |
163.18 | |
163.18 |
Safety and Handling
GHS H Statement:
May cause an allergic skin reaction.
May cause damage to organs through prolonged or repeated exposure.
Suspected of damaging fertility.
GHS P Statement:
Wear eye protection/face protection.
Use personal protective equipment as required.
Do not breathe dust/fume/gas/mist/vapors/spray.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
TSCA : TSCA
RUO â Research Use Only