Please login to your online account to display your discounted pricing

(S)-(-)-Indoline-2-carboxylic acid, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material (S)-(-)-Indoline-2-carboxylic acid
Name Note 98%
CAS 79815-20-6
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C9H9NO2
Formula Weight 163.18
MDL Number MFCD00070578
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC342080050

  SDS
thermo fisher scientific
342080050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Powder
Formula Weight 163.18
Quantity 5g
Color Beige to brown
TSCA TSCA
Specific Rotation Condition −112.5° (20°C c=1,1N HCl)
CAS Max % 100.0
Specific Rotation -108° to -117° (20°C, 589nm) (c=1, 1N HCl)
CAS Min % 97.5
Health Hazard 3 GHS P Statement:
Wear eye protection/face protection.
Use personal protective equipment as required.
Do not breathe dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material (S)-(-)-Indoline-2-carboxylic acid
Infrared Spectrum Authentic
Packaging Glass bottle
Name Note 98%
Assay Percent Range 98%
Molecular Formula C9H9NO2
MDL Number MFCD00070578
CAS 79815-20-6
Health Hazard 2 GHS H Statement:
May cause an allergic skin reaction.
May cause damage to organs through prolonged or repeated exposure.
Suspected of damaging fertility.
Safety and Handling

GHS P Statement:
Wear eye protection/face protection.
Use personal protective equipment as required.
Do not breathe dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause an allergic skin reaction.
May cause damage to organs through prolonged or repeated exposure.
Suspected of damaging fertility.