missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ S-Hydroxymethyl thiobenzoate, 98%

$70.16 - $231.32

Chemical Identifiers

CAS 23853-33-0
Molecular Formula C8H8O2S
Molecular Weight (g/mol) 168.21
MDL Number MFCD00014423
InChI Key MNTZJQAAOYSCMM-UHFFFAOYSA-N
Synonym s-hydroxymethyl benzenecarbothioate, s-hydroxymethyl benzothioate, s-hydroxymethyl thiobenzoate, hydroxymethylthio phenyl ketone, hydroxymethyl sulfanyl phenyl methanone, benzoyl-oxymethyl-thiamine, s-hydroxymethylthiobenzoate, hydroxymethylbenzoyl sulfide
PubChem CID 287157
IUPAC Name S-(hydroxymethyl) benzenecarbothioate
SMILES C1=CC=C(C=C1)C(=O)SCO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0052306 View Documents Alfa Aesar™
L0052306
5g Each for $70.16
 
AAL0052314 View Documents Alfa Aesar™
L0052314
25g Each for $231.32
 
Specifications

Chemical Identifiers

23853-33-0
168.21
MNTZJQAAOYSCMM-UHFFFAOYSA-N
287157
C1=CC=C(C=C1)C(=O)SCO
C8H8O2S
MFCD00014423
s-hydroxymethyl benzenecarbothioate, s-hydroxymethyl benzothioate, s-hydroxymethyl thiobenzoate, hydroxymethylthio phenyl ketone, hydroxymethyl sulfanyl phenyl methanone, benzoyl-oxymethyl-thiamine, s-hydroxymethylthiobenzoate, hydroxymethylbenzoyl sulfide
S-(hydroxymethyl) benzenecarbothioate

Specifications

Unpleasant
5g
C8H8O2S
2574909
MNTZJQAAOYSCMM-UHFFFAOYSA-N
S-(hydroxymethyl) benzenecarbothioate
287157
98%
45°C to 48°C
23853-33-0
MFCD00014423
s-hydroxymethyl benzenecarbothioate, s-hydroxymethyl benzothioate, s-hydroxymethyl thiobenzoate, hydroxymethylthio phenyl ketone, hydroxymethyl sulfanyl phenyl methanone, benzoyl-oxymethyl-thiamine, s-hydroxymethylthiobenzoate, hydroxymethylbenzoyl sulfide
C1=CC=C(C=C1)C(=O)SCO
168.21
168.21
S-Hydroxymethyl thiobenzoate