Please login to your online account to display your discounted pricing

(S)-(+)-γ-Hydroxymethyl-γ-butyrolactone, 97%, ACROS Organics™


Quantity 250mg
Packaging Glass Bottle

Manufacturer: thermo fisher scientific  AC337642500

 View more versions of this product

Catalog No. AC337642500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 116.12
Quantity 250mg
Packaging Glass Bottle
Density 1.23g/mL
Specific Rotation Condition +33.5° (20°C 589nm,c=3,EtOH)
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
MDL Number MFCD00066224
CAS 32780-06-6
Chemical Name or Material (S)-(+)-γ-Hydroxymethyl-γ-butyrolactone
Molecular Formula C5H8O3
Synonym (S)-(+)-Dihydro-5-(hydroxymethyl)-2(3H)-furanone
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material (S)-(+)-γ-Hydroxymethyl-γ-butyrolactone
CAS 32780-06-6
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C5H8O3
Formula Weight 116.12
MDL Number MFCD00066224
Synonym (S)-(+)-Dihydro-5-(hydroxymethyl)-2(3H)-furanone
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

SDS & Safety Documents