missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(S)-(+)-γ-Hydroxymethyl-γ-butyrolactone, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 337640010

 View more versions of this product

Catalog No. AC337640010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

(S)-(+)-γ-Hydroxymethyl-γ-butyrolactone
96.0
1.2300g/mL
>110°C
Glass bottle
1.4680 to 1.4720
1g
+33.5° (20°C 589nm,c=3,EtOH)
s-5-hydroxymethyl dihydrofuran-2 3h-one, s-+-dihydro-5-hydroxymethyl-2 3h-furanone, s-+-dihydro-5-hydroxymethyl furanone, s-+-gamma-hydroxymethyl-gamma-butyrolactone, 5s-5-hydroxymethyl oxolan-2-one, s-5-hydroxymethyldihydrofuran-2-one, s-5-hydroxymethyl-dihydrofuran-2 3h-one, 2 3h-furanone, dihydro-5-hydroxymethyl-, 5s, s-4-hydroxymethyl butyrolactone, s-+-2',3'-dideoxyribonolactone
NSISJFFVIMQBRN-BYPYZUCNSA-N
(5S)-5-(hydroxymethyl)oxolan-2-one
2733813
97%
32780-06-6
100.0
110°C to 115°C (2.0mmHg)
96% min. (GC)
C5H8O3
MFCD00066224
1.23
+ 33.50
Solubility in water: miscible.
C1CC(=O)OC1CO
116.12
116.12

Chemical Identifiers

32780-06-6
116.12
NSISJFFVIMQBRN-BYPYZUCNSA-N
2733813
C1CC(=O)OC1CO
C5H8O3
MFCD00066224
s-5-hydroxymethyl dihydrofuran-2 3h-one, s-+-dihydro-5-hydroxymethyl-2 3h-furanone, s-+-dihydro-5-hydroxymethyl furanone, s-+-gamma-hydroxymethyl-gamma-butyrolactone, 5s-5-hydroxymethyl oxolan-2-one, s-5-hydroxymethyldihydrofuran-2-one, s-5-hydroxymethyl-dihydrofuran-2 3h-one, 2 3h-furanone, dihydro-5-hydroxymethyl-, 5s, s-4-hydroxymethyl butyrolactone, s-+-2',3'-dideoxyribonolactone
(5S)-5-(hydroxymethyl)oxolan-2-one
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Avoid breathing dust/fume/

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only