Please login to your online account to display your discounted pricing

(S)-(+)-Glycidyl nosylate, 98%, ACROS Organics™


Quantity 10g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC296420100

 View more versions of this product

Catalog No. AC296420100

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Needles or Powder
Assay ≥97.5% (GC)
Formula Weight 259.23
Quantity 10g
Color Off-white to yellow-brown
Melting Point 61° to 65°C
Specific Rotation Condition + 21.50 (20.00°C c=2,CHCl3)
Chemical Name or Material (S)-(+)-Glycidyl nosylate
Assay Percent Range 98%
Molecular Formula C9H9NO6S
CAS 115314-14-2
Infrared Spectrum Authentic
Packaging Glass bottle
Specific Rotation (20°C, 589nm) (c=2, CHCl3) > +21.5°

Specifications

Chemical Name or Material (S)-(+)-Glycidyl nosylate
Assay ≥97.5% (GC)
Assay Percent Range 98%
Molecular Formula C9H9NO6S
Formula Weight 259.23