missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
(S)-Glycidyl Butyrate 98.0+%, TCI America™
$165.73 - $536.30
Chemical Identifiers
CAS | 65031-96-1 |
---|---|
Molecular Formula | C7H12O3 |
Molecular Weight (g/mol) | 144.17 |
MDL Number | MFCD00273369 |
InChI Key | YLNSNVGRSIOCEU-LURJTMIESA-N |
Synonym | 2s-oxiran-2-yl methyl butanoate, butanoic acid, 2s-oxiranylmethyl ester, butyric acid s-glycidyl ester, pubchem5837, s-+-glycidyl butyrate, s-2-butyryloxymethyl oxirane, s-glycidyl butanoate, s-glycidyl butyrate, s-oxiran-2-ylmethyl butyrate |
PubChem CID | 6999819 |
IUPAC Name | [(2S)-oxiran-2-yl]methyl butanoate |
SMILES | CCCC(=O)OCC1CO1 |
Chemical Identifiers
65031-96-1 | |
144.17 | |
YLNSNVGRSIOCEU-LURJTMIESA-N | |
6999819 | |
CCCC(=O)OCC1CO1 |
C7H12O3 | |
MFCD00273369 | |
2s-oxiran-2-yl methyl butanoate, butanoic acid, 2s-oxiranylmethyl ester, butyric acid s-glycidyl ester, pubchem5837, s-+-glycidyl butyrate, s-2-butyryloxymethyl oxirane, s-glycidyl butanoate, s-glycidyl butyrate, s-oxiran-2-ylmethyl butyrate | |
[(2S)-oxiran-2-yl]methyl butanoate |
Specifications
90°C | |
5g | |
C7H12O3 | |
2s-oxiran-2-yl methyl butanoate, butanoic acid, 2s-oxiranylmethyl ester, butyric acid s-glycidyl ester, pubchem5837, s-+-glycidyl butyrate, s-2-butyryloxymethyl oxirane, s-glycidyl butanoate, s-glycidyl butyrate, s-oxiran-2-ylmethyl butyrate | |
CCCC(=O)OCC1CO1 | |
144.17 | |
144.17 | |
≥98.0% (GC) |
Yellow | |
65031-96-1 | |
MFCD00273369 | |
YLNSNVGRSIOCEU-LURJTMIESA-N | |
[(2S)-oxiran-2-yl]methyl butanoate | |
6999819 | |
Clear Liquid at 20°C | |
(S)-Glycidyl Butyrate |
Safety and Handling
TSCA : No