Please login to your online account to display your discounted pricing

Alfa Aesar™ (S)-(+)-Glycidyl butyrate, 98%, ee 98%

$44.85 - $44.85

Chemical Identifiers

Chemical Name or Material (S)-(+)-Glycidyl butyrate
CAS 65031-96-1
Molecular Formula C7H12O3
Formula Weight 144.17g/mol
MDL Number MFCD00273369
Beilstein 8198560
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1855803

 
alfa aesar™
L18558-03
1g Each for $44.85
Description & Specifications

Specifications

Boiling Point 90°C (19mmHg)
Quantity 1g
Flash Point 85°C (185°F)
Optical Rotation +31° (Neat)
Chemical Name or Material (S)-(+)-Glycidyl butyrate
CAS 65031-96-1
Assay 98%
Assay Percent Notes ee 98%
Molecular Formula C7H12O3
Formula Weight 144.17g/mol
MDL Number MFCD00273369
Beilstein 8198560
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H312-H315-H319-H335-H227
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Health Hazard 3 GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H312-H315-H319-H335-H227
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.