Please login to your online account to display your discounted pricing

(S)-(-)-Glycidol, 97%, (98% ee), ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC411000010

 View more versions of this product

Catalog No. AC411000010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 74.08
Physical Form Liquid
Assay ≥96.0% (GC)
Quantity 1g
Color Clear colorless to light yellow
Density 1.116g/mL
CAS Max % 100.0
CAS Min % 96.0
Enantiomeric Excess 97.5% min. (GC)
Packaging Glass bottle
Specific Rotation −12° to −18° (20°C, 589nm) (neat)
Health Hazard 3 GHS P Statement:
IF exposed or concerned: Get medical advice/attention.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Suspected of causing genetic defects.
May cause cancer.
May damage fertility.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.
Toxic if inhaled.
Name Note 98% ee
CAS 60456-23-7
Molecular Formula C3H6O2
Chemical Name or Material (S)-(-)-Glycidol
MDL Number MFCD00074874
Assay Percent Range 97%
Refractive Index 1.4310 to 1.4340 (20°C, 589nm)
Infrared Spectrum Authentic
Synonym (S)-2, 3-Epoxy-1-propanol
Health Hazard 1 GHS Signal Word: Danger

Specifications

Chemical Name or Material (S)-(-)-Glycidol
Name Note 98% ee
CAS 60456-23-7
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0% (GC)
Assay Percent Range 97%
Molecular Formula C3H6O2
Formula Weight 74.08
MDL Number MFCD00074874
Synonym (S)-2, 3-Epoxy-1-propanol
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF exposed or concerned: Get medical advice/attention.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Suspected of causing genetic defects.
May cause cancer.
May damage fertility.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.
Toxic if inhaled.

SDS & Safety Documents