Please login to your online account to display your discounted pricing

Alfa Aesar™ S-Furfuryl thiopropionate, 99%

$38.18 - $481.80

Chemical Identifiers

Chemical Name or Material S-Furfuryl thiopropionate
CAS 59020-85-8
Molecular Formula C8H10O2S
Formula Weight 170.23
MDL Number MFCD00040264
Synonym Thiopropionic acid S-furfuryl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2235106

 
alfa aesar™
B2235106
5g Each for $38.18

AAB2235122

 
alfa aesar™
B2235122
100g Each for $481.80
Description & Specifications

Specifications

Formula Weight 170.23
Chemical Name or Material S-Furfuryl thiopropionate
TSCA TSCA
Assay 99%
Molecular Formula C8H10O2S
MDL Number MFCD00040264
CAS 59020-85-8
Synonym Thiopropionic acid S-furfuryl ester
Boiling Point 219° to 221°C
Quantity 5g
Flash Point 97°C (207°F)