Please login to your online account to display your discounted pricing

Alfa Aesar™ S-Furfuryl thioacetate, 99%

Chemical Identifiers

Chemical Name or Material S-Furfuryl thioacetate
CAS 13678-68-7
Molecular Formula C7H8O2S
Formula Weight 156.2
MDL Number MFCD00010083
UN Number UN3334
Synonym Thioacetic acid S-furfuryl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2182106

  SDS
alfa aesar™
B2182106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2182114

  SDS
alfa aesar™
B2182114
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 156.2
Chemical Name or Material S-Furfuryl thioacetate
TSCA TSCA
Assay 99%
Molecular Formula C7H8O2S
MDL Number MFCD00010083
CAS 13678-68-7
UN Number UN3334
Synonym Thioacetic acid S-furfuryl ester
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Health Hazard 3 GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Boiling Point 211° to 212°C
Quantity 5g
Flash Point 92°C (199°F)
Odor Stench
DOT Information Transport Hazard Class: 9; Proper Shipping Name: AVIATION REGULATED LIQUID, N.O.S.
Safety and Handling

Warning

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.