missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
S-Ethylisothiourea Hydrobromide 98.0+%, TCI America™
Supplier: TCI America E0182100G
Specifications
S-Ethylisothiourea Hydrobromide | |
90°C | |
C3H9BrN2S | |
100 g | |
SWXXKWPYNMZFTE-UHFFFAOYSA-N | |
ethyl carbamimidothioate;hydrobromide | |
200213 | |
≥98.0% (T) |
1071-37-0 | |
White-Yellow | |
MFCD00012585 | |
s-ethylisothiourea hydrobromide, 2-ethyl-2-thiopseudourea hydrobromide, ethiron, ethiron bromide, s-ethylthiuronium bromide, s-ethylisothiouronium hydrobromide, bromide s-ethylisothiuronium, s-ethylisothiuronium bromide, usaf el-18, isothuron hydrobromide | |
CCSC(=N)N.Br | |
185.083 | |
185.08 | |
Crystalline Powder |
Chemical Identifiers
1071-37-0 | |
185.083 | |
SWXXKWPYNMZFTE-UHFFFAOYSA-N | |
200213 | |
CCSC(=N)N.Br |
C3H9BrN2S | |
MFCD00012585 | |
s-ethylisothiourea hydrobromide, 2-ethyl-2-thiopseudourea hydrobromide, ethiron, ethiron bromide, s-ethylthiuronium bromide, s-ethylisothiouronium hydrobromide, bromide s-ethylisothiuronium, s-ethylisothiuronium bromide, usaf el-18, isothuron hydrobromide | |
ethyl carbamimidothioate;hydrobromide |
Safety and Handling
EINECSNumber : (2)-2866
RTECSNumber : UM6825000
TSCA : No