Please login to your online account to display your discounted pricing

Alfa Aesar™ S-Ethyl thiopropionate, 97%

Chemical Identifiers

Chemical Name or Material S-Ethyl thiopropionate
CAS 2432-42-0
Molecular Formula C5H10OS
Formula Weight 118.2
MDL Number MFCD00027016
UN Number UN1993
Beilstein 1740740
Synonym Thiopropionic acid S-ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2491806

  SDS
alfa aesar™
B2491806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2491814

  SDS
alfa aesar™
B2491814
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2491822

  SDS
alfa aesar™
B2491822
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 118.2
MDL Number MFCD00027016
Flash Point 27°C (80°F)
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
Quantity 5g
Health Hazard 1 Warning
Chemical Name or Material S-Ethyl thiopropionate
Health Hazard 2 GHS H Statement
H226
Flammable liquid and vapour.
Odor Stench
Molecular Formula C5H10OS
Boiling Point 137° to 138°C
Assay 97%
Beilstein 1740740
UN Number UN1993
CAS 2432-42-0
Synonym Thiopropionic acid S-ethyl ester
Health Hazard 3 GHS P Statement
P210-P261
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Safety and Handling

Warning

GHS P Statement
P210-P261
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.

GHS H Statement
H226
Flammable liquid and vapour.