Learn More
S-Ethyl thioacetate, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1068006
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
S-Ethyl thioacetate | |
0.98 | |
20°C (68°F) | |
C4H8OS | |
MFCD00015178 | |
UN1993 | |
s-ethyl thioacetate, ethanethioic acid, ethyl ester, ethanethioic acid, s-ethyl ester, ethyl thiolacetate, ethyl ethanethioate, ethanethioic acid s-ethyl ester, s-ethyl thiolacetate, acetic acid, thio-, ethyl ester, thioethyl compound, acetic acid, thio-, s-ethyl ester | |
APTGPWJUOYMUCE-UHFFFAOYSA-N | |
S-ethyl ethanethioate | |
61171 | |
≥98% |
625-60-5 | |
116°C to 117°C | |
Stench | |
1.46 | |
5 g | |
1737643 | |
Insoluble in water | |
CCSC(=O)C | |
104.167 | |
104.18 |
Chemical Identifiers
625-60-5 | |
104.167 | |
APTGPWJUOYMUCE-UHFFFAOYSA-N | |
61171 | |
CCSC(=O)C |
C4H8OS | |
MFCD00015178 | |
s-ethyl thioacetate, ethanethioic acid, ethyl ester, ethanethioic acid, s-ethyl ester, ethyl thiolacetate, ethyl ethanethioate, ethanethioic acid s-ethyl ester, s-ethyl thiolacetate, acetic acid, thio-, ethyl ester, thioethyl compound, acetic acid, thio-, s-ethyl ester | |
S-ethyl ethanethioate |
Safety and Handling
GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H225-H315-H319-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber : 210-904-9
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only