missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-(-)-DM-SEGPHOS(regR) 95.0+%, TCI America™
$106.25 - $313.16
Chemical Identifiers
| CAS | 210169-57-6 |
|---|---|
| Molecular Formula | C46H44O4P2 |
| Molecular Weight (g/mol) | 722.802 |
| MDL Number | MFCD09753008 |
| InChI Key | XJJVPYMFHXMROQ-UHFFFAOYSA-N |
| Synonym | (S)-(-)-5,5′C-Bis[di(3,5-xylyl)phosphino]-4,4′C-bi-1,3-benzodioxole, (S)-(-)-4,4′C-Bis[di(3,5-xylyl)phosphino]-3,3′C-bi(1,2-methylenedioxybenzene) |
| PubChem CID | 11297101 |
| IUPAC Name | [4-[5-bis(3,5-dimethylphenyl)phosphanyl-1,3-benzodioxol-4-yl]-1,3-benzodioxol-5-yl]-bis(3,5-dimethylphenyl)phosphane |
| SMILES | CC1=CC(=CC(=C1)P(C2=C(C3=C(C=C2)OCO3)C4=C(C=CC5=C4OCO5)P(C6=CC(=CC(=C6)C)C)C7=CC(=CC(=C7)C)C)C8=CC(=CC(=C8)C)C)C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
D4498200MG
|
TCI America
D4498200MG |
200 mg |
Each for $106.25
|
|
|||||
|
D44981G
|
TCI America
D44981G |
1 g |
Each for $313.16
|
|
|||||
Chemical Identifiers
| 210169-57-6 | |
| 722.802 | |
| XJJVPYMFHXMROQ-UHFFFAOYSA-N | |
| 11297101 | |
| CC1=CC(=CC(=C1)P(C2=C(C3=C(C=C2)OCO3)C4=C(C=CC5=C4OCO5)P(C6=CC(=CC(=C6)C)C)C7=CC(=CC(=C7)C)C)C8=CC(=CC(=C8)C)C)C |
| C46H44O4P2 | |
| MFCD09753008 | |
| (S)-(-)-5,5′C-Bis[di(3,5-xylyl)phosphino]-4,4′C-bi-1,3-benzodioxole, (S)-(-)-4,4′C-Bis[di(3,5-xylyl)phosphino]-3,3′C-bi(1,2-methylenedioxybenzene) | |
| [4-[5-bis(3,5-dimethylphenyl)phosphanyl-1,3-benzodioxol-4-yl]-1,3-benzodioxol-5-yl]-bis(3,5-dimethylphenyl)phosphane |
Specifications
| 210169-57-6 | |
| C46H44O4P2 | |
| 200 mg | |
| XJJVPYMFHXMROQ-UHFFFAOYSA-N | |
| [4-[5-bis(3,5-dimethylphenyl)phosphanyl-1,3-benzodioxol-4-yl]-1,3-benzodioxol-5-yl]-bis(3,5-dimethylphenyl)phosphane | |
| 11297101 | |
| ≥95.0% (HPLC) | |
| (S)-(-)-DM-SEGPHOS(regR) |
| White-Yellow | |
| MFCD09753008 | |
| (S)-(-)-5,5′C-Bis[di(3,5-xylyl)phosphino]-4,4′C-bi-1,3-benzodioxole, (S)-(-)-4,4′C-Bis[di(3,5-xylyl)phosphino]-3,3′C-bi(1,2-methylenedioxybenzene) | |
| CC1=CC(=CC(=C1)P(C2=C(C3=C(C=C2)OCO3)C4=C(C=CC5=C4OCO5)P(C6=CC(=CC(=C6)C)C)C7=CC(=CC(=C7)C)C)C8=CC(=CC(=C8)C)C)C | |
| 722.802 | |
| 722.80 | |
| Crystalline Powder |
Safety and Handling
TSCA : No