Please login to your online account to display your discounted pricing

s-Diphenylcarbazone, ACS Grade, LabChem™

$123.57 - $123.57

Chemical Identifiers

Chemical Name or Material s-Diphenylcarbazone
CAS 538-62-5
CAS Max % 51
CAS 140-22-7
CAS Max % 49
Purity Grade ACS Grade
Linear Formula C13H14N4O
Formula Weight 242.28
UN Number UN1325
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC136757

 
lab chem
LC136757
Amber Glass 10g Each for $123.57
Description & Specifications

Specifications

Formula Weight 242.28
Chemical Name or Material s-Diphenylcarbazone
Identification Passes Test
Packaging Amber Glass
Purity Grade ACS Grade
Physical Form Powder
Sensitivity Passes Test
Linear Formula C13H14N4O
Solubility Soluble in water
CAS 538-62-5
Storage Information Room Temperature
UN Number UN1325
Health Hazard 1 Warning
Color Orange
Quantity 10g
CAS Max % 51
Health Hazard 2 GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
CAS 140-22-7
Health Hazard 3 GHS P Statement
Avoid breathing dust.
Use only outdoors or in a well-ventilated area.
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
Take off contaminated clothing and wash before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor if you feel unwell.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Store locked up in a well-ventilated place.
Keep container tightly closed.
Dispose of contents/container to comply with local, state and federal regulations.
Decomposition Information Fume; Carbon monoxide; Carbon dioxide
CAS Max % 49
Safety and Handling

Warning

GHS P Statement
Avoid breathing dust.
Use only outdoors or in a well-ventilated area.
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
Take off contaminated clothing and wash before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor if you feel unwell.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Store locked up in a well-ventilated place.
Keep container tightly closed.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.