missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

S-Curve™ WD Series Wiper Dispenser

Unique design maintains cleanliness

$149.67 - $149.67

Specifications

Color Clear
Product Type Wiper Dispenser
Edges Flame polished
For Use With (Application) Offers quick and easy access
For Use With (Equipment) For wipes, table top use only
View More Specs

Products 1
Catalog Number Mfr. No. Depth (English) Depth (Metric) Width (English) Width (Metric) Price Quantity  
Catalog Number Mfr. No. Depth (English) Depth (Metric) Width (English) Width (Metric) Price Quantity  
19-034-273 View Documents S-Curve™
WD95
9.5 in. 22.9cm 17.5 in. 15.8cm Each for $149.67
Description

Description

  • Offers quick and easy access
  • 0.25 in.-thick (0.6cm) clear acrylic construction
  • Flame-treated edges for smoothness
  • Cover helps protect wipers
  • Two models hold different wiper sizes: 9 x 9 (22.9 x 22.9cm) or 12 x 12 in. (30.5 x 30.5cm)

Specifications

Specifications

Clear
Flame polished
For wipes, table top use only
7.6cm
Acrylic
Wiper Dispenser
Offers quick and easy access
3 in.
Rubber pads on bottom corners
0.25 in.