S-Curve™ Low-Profile Glove Dispenser with 5 Compartments
Dispenser features a single, sloping lid with full-length hinge to maintain HEPA flow. Top loading with front access holes for convenient access to products.
Supplier: S-Curve™ AT105
Description
- Store and organize multiple sizes of gloves in a low-profile design
- Clear material allows for constant visibility of stock to ensure that gloves never run out
- Single, sloping lid with full-length hinge to maintain HEPA flow
- Flame-treated edges ensure a smooth finish
- Bracket included for wall mounting
Specifications
Glove Dispenser | |
6 x 30 x 16 in. | |
Acrylic | |
Top Loading |
Clear | |
Gloves | |
5 | |
Wall Bracket |