Please login to your online account to display your discounted pricing

(S)-CPW 399, Tocris

Subtype-selective weakly desensitizing AMPA agonist

$244.81 - $997.23

Chemical Identifiers

Chemical Name or Material (S)-CPW 399
Assay Percent Range >98%
Molecular Formula C10H13N3O4
Formula Weight 239.23
Synonym (S)-α-Amino-2,3,4,5,6,7-hexahydro-2,4-dioxo-1H-cyclopentapyrimidine-1-propanoic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

154310

 
r & d systems
154310
10mg Each for $244.81

154350

 
r & d systems
154350
50mg Each for $997.23
Description & Specifications

Specifications

Formula Weight 239.23
Quantity 10mg
Chemical Name or Material (S)-CPW 399
Assay Percent Range >98%
Molecular Formula C10H13N3O4
CAS 389888-02-2
Synonym (S)-α-Amino-2,3,4,5,6,7-hexahydro-2,4-dioxo-1H-cyclopentapyrimidine-1-propanoic acid
Recommended Storage Desiccate at -20°C
Safety and Handling

Desiccate at -20°C