Please login to your online account to display your discounted pricing

S-Butyrylthiocholine iodide, 98%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  108150250

 View more versions of this product

Catalog No. AC108150250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material S-Butyrylthiocholine iodide
CAS 1866-16-6
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
Causes serious eye irritation. Causes skin irritation. May cause respiratory irritation.
Health Hazard 3 GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Infrared Spectrum Authentic
Physical Form Crystals or Crystalline Powder
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C9H20INOS
Linear Formula CH3CH2CH2COSCH2CH2N(CH3)3I
Packaging Glass bottle
Formula Weight 317.22
Synonym (2-Mercaptoethyl)trimethylammonium iodide butyrate
Melting Point 172° to 176°C
Color White
Quantity 25g

Specifications

Chemical Name or Material S-Butyrylthiocholine iodide
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C9H20INOS
Linear Formula CH3CH2CH2COSCH2CH2N(CH3)3I
Formula Weight 317.22
Synonym (2-Mercaptoethyl)trimethylammonium iodide butyrate
Accessories

Warning

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
Causes serious eye irritation. Causes skin irritation. May cause respiratory irritation.

SDS & Safety Documents