Please login to your online account to display your discounted pricing

(S)-BOC-2,4-Dichlorophenylalanine, 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material (S)-BOC-2, 4-Dichlorophenylalanine
CAS 114873-04-0
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C14 H17 Cl2 N O4
Formula Weight 334.2
MDL Number MFCD01862942
Synonym N-tert-Butoxycarbonyl-2, 4-dichloro-L-phenylalanine
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC368820050

  SDS
fisher scientific acros
AC368820050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5%
Formula Weight 334.2
Physical Form Powder
Quantity 5g
Color White
CAS Max % 100.0
Specific Rotation -30° to -34° (20°C, 589nm) (C=1, Methanol)
Assay Percent Range 99%
Molecular Formula C14 H17 Cl2 N O4
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS Min % 98.5
Synonym N-tert-Butoxycarbonyl-2, 4-dichloro-L-phenylalanine
Infrared Spectrum Authentic
Chemical Name or Material (S)-BOC-2, 4-Dichlorophenylalanine
CAS 114873-04-0
Packaging Glass bottle
MDL Number MFCD01862942
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.