missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(S)-(-)-beta-Methylphenethylamine, TCI America™

$197.09 - $673.77

Chemical Identifiers

CAS 17596-79-1
Molecular Formula C9H13N
Molecular Weight (g/mol) 135.21
MDL Number MFCD00216741
InChI Key AXORVIZLPOGIRG-MRVPVSSYSA-N
Synonym (S)-(-)-2-Phenyl-1-propylamine
PubChem CID 1547950
IUPAC Name (2S)-2-phenylpropan-1-amine
SMILES CC(CN)C1=CC=CC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M17475G View Documents TCI America
M17475G
5g Each for $197.09
 
M174725G View Documents TCI America
M174725G
25g Each for $673.77
 
Specifications

Chemical Identifiers

17596-79-1
135.21
AXORVIZLPOGIRG-MRVPVSSYSA-N
1547950
CC(CN)C1=CC=CC=C1
C9H13N
MFCD00216741
(S)-(-)-2-Phenyl-1-propylamine
(2S)-2-phenylpropan-1-amine

Specifications

Yellow
17596-79-1
MFCD00216741
(S)-(-)-2-Phenyl-1-propylamine
CC(CN)C1=CC=CC=C1
135.21
135.21
≥98.0% (GC,T)
5g
C9H13N
2735
AXORVIZLPOGIRG-MRVPVSSYSA-N
(2S)-2-phenylpropan-1-amine
1547950
Clear Liquid at 20°C
(S)-(-)-beta-Methylphenethylamine
Safety and Handling

Safety and Handling

TSCA : No