missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
(S)-beta-(Carbobenzoxyamino)-gamma-butyrolactone 98.0+%, TCI America™
$59.42 - $178.25
Chemical Identifiers
CAS | 87219-29-2 |
---|---|
Molecular Formula | C12H13NO4 |
Molecular Weight (g/mol) | 235.239 |
MDL Number | MFCD00216569 |
InChI Key | BNIBNUOPVTZWRT-JTQLQIEISA-N |
Synonym | benzyl 3s-5-oxotetrahydrofuran-3-ylcarbamate, benzyl n-3s-5-oxooxolan-3-yl carbamate, benzyl s---tetrahydro-5-oxo-3-furanyl-carbama, benzyl s---tetrahydro-5-oxo-3-furanylcarbamate, benzyl s-tetrahydro-5-oxo-3-furanylcarbamate, carbamic acid, 3s-tetrahydro-5-oxo-3-furanyl-, phenylmethyl ester, carbamic acid, 3s-tetrahydro-5-oxo-3-furanyl-, phenylmethyl ester 9ci, n-3s-5-oxo 3-2,3,4-trihydrofuryl phenylmethoxy carboxamide, s-benzyl 5-oxotetrahydrofuran-3-yl carbamate, s-benzyl-5-oxo-tetrahydro-furan-3-ylcarbamate |
PubChem CID | 697922 |
IUPAC Name | benzyl N-[(3S)-5-oxooxolan-3-yl]carbamate |
SMILES | C1C(COC1=O)NC(=O)OCC2=CC=CC=C2 |
Chemical Identifiers
87219-29-2 | |
235.239 | |
BNIBNUOPVTZWRT-JTQLQIEISA-N | |
697922 | |
C1C(COC1=O)NC(=O)OCC2=CC=CC=C2 |
C12H13NO4 | |
MFCD00216569 | |
benzyl 3s-5-oxotetrahydrofuran-3-ylcarbamate, benzyl n-3s-5-oxooxolan-3-yl carbamate, benzyl s---tetrahydro-5-oxo-3-furanyl-carbama, benzyl s---tetrahydro-5-oxo-3-furanylcarbamate, benzyl s-tetrahydro-5-oxo-3-furanylcarbamate, carbamic acid, 3s-tetrahydro-5-oxo-3-furanyl-, phenylmethyl ester, carbamic acid, 3s-tetrahydro-5-oxo-3-furanyl-, phenylmethyl ester 9ci, n-3s-5-oxo 3-2,3,4-trihydrofuryl phenylmethoxy carboxamide, s-benzyl 5-oxotetrahydrofuran-3-yl carbamate, s-benzyl-5-oxo-tetrahydro-furan-3-ylcarbamate | |
benzyl N-[(3S)-5-oxooxolan-3-yl]carbamate |
Specifications
White | |
1g | |
C12H13NO4 | |
benzyl 3s-5-oxotetrahydrofuran-3-ylcarbamate, benzyl n-3s-5-oxooxolan-3-yl carbamate, benzyl s---tetrahydro-5-oxo-3-furanyl-carbama, benzyl s---tetrahydro-5-oxo-3-furanylcarbamate, benzyl s-tetrahydro-5-oxo-3-furanylcarbamate, carbamic acid, 3s-tetrahydro-5-oxo-3-furanyl-, phenylmethyl ester, carbamic acid, 3s-tetrahydro-5-oxo-3-furanyl-, phenylmethyl ester 9ci, n-3s-5-oxo 3-2,3,4-trihydrofuryl phenylmethoxy carboxamide, s-benzyl 5-oxotetrahydrofuran-3-yl carbamate, s-benzyl-5-oxo-tetrahydro-furan-3-ylcarbamate | |
C1C(COC1=O)NC(=O)OCC2=CC=CC=C2 | |
235.239 | |
235.24 | |
≥98.0% (HPLC,N) |
106°C | |
87219-29-2 | |
MFCD00216569 | |
BNIBNUOPVTZWRT-JTQLQIEISA-N | |
benzyl N-[(3S)-5-oxooxolan-3-yl]carbamate | |
697922 | |
Crystal-Powder at 20°C | |
(S)-beta-(Carbobenzoxyamino)-gamma-butyrolactone |
Safety and Handling
TSCA : No