missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

S-Benzyl-L-cysteine, TCI America™

$19.86 - $103.89

Chemical Identifiers

CAS 3054-01-1
Molecular Formula C10H13NO2S
Molecular Weight (g/mol) 211.28
MDL Number MFCD00002613
InChI Key GHBAYRBVXCRIHT-UHFFFAOYNA-N
Synonym s-benzyl-l-cysteine, benzylcysteine, h-cys bzl-oh, s-benzylcysteine, poly-s-benzylcysteine, r-2-amino-3-benzylthio propanoic acid, cysteine, s-phenylmethyl, chembl63130, unii-9vre13m548, 3-benzylthioalanine
PubChem CID 193613
ChEBI CHEBI:41085
IUPAC Name (2R)-2-amino-3-benzylsulfanylpropanoic acid
SMILES C1=CC=C(C=C1)CSCC(C(=O)O)N
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B08651G View Documents TCI America
B08651G
1g Each for $19.86
 
B086525G View Documents TCI America
B086525G
25g Each for $103.89
 
B08655G View Documents TCI America
B08655G
5g Each for $33.61
 
Specifications

Chemical Identifiers

3054-01-1
211.28
GHBAYRBVXCRIHT-UHFFFAOYNA-N
193613
(2R)-2-amino-3-benzylsulfanylpropanoic acid
C10H13NO2S
MFCD00002613
s-benzyl-l-cysteine, benzylcysteine, h-cys bzl-oh, s-benzylcysteine, poly-s-benzylcysteine, r-2-amino-3-benzylthio propanoic acid, cysteine, s-phenylmethyl, chembl63130, unii-9vre13m548, 3-benzylthioalanine
CHEBI:41085
C1=CC=C(C=C1)CSCC(C(=O)O)N

Specifications

White
3054-01-1
MFCD00002613
GHBAYRBVXCRIHT-UHFFFAOYNA-N
(2R)-2-amino-3-benzylsulfanylpropanoic acid
193613
211.28
≥98.0% (HPLC,T)
1g
C10H13NO2S
s-benzyl-l-cysteine, benzylcysteine, h-cys bzl-oh, s-benzylcysteine, poly-s-benzylcysteine, r-2-amino-3-benzylthio propanoic acid, cysteine, s-phenylmethyl, chembl63130, unii-9vre13m548, 3-benzylthioalanine
C1=CC=C(C=C1)CSCC(C(=O)O)N
211.28
CHEBI:41085
Crystal-Powder at 20°C
S-Benzyl-L-cysteine
Safety and Handling

Safety and Handling

EINECSNumber : (9)-1591

TSCA : Yes