missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ S-Benzhydryl-N-Boc-L-cysteine, 98%

$30.07 - $351.61

Chemical Identifiers

CAS 21947-97-7
Molecular Formula C21H25NO4S
Molecular Weight (g/mol) 387.494
MDL Number MFCD00151869
InChI Key AMTHPOXIVDHMLZ-KRWDZBQOSA-N
Synonym boc-cys dpm-oh, boc-s-diphenylmethyl-l-cysteine, 2r-2-tert-butoxycarbonyl amino-3-diphenylmethyl sulfanyl propanoic acid, l-cysteine,n-1,1-dimethylethoxy carbonyl-s-diphenylmethyl
PubChem CID 7015794
IUPAC Name (2R)-3-benzhydrylsulfanyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
SMILES CC(C)(C)OC(=O)NC(CSC(C1=CC=CC=C1)C2=CC=CC=C2)C(=O)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH66001MD View Documents Alfa Aesar™
H66001MD
250mg Each for $30.07
 
AAH6600103 View Documents Alfa Aesar™
H6600103
1g Each for $88.68
 
AAH6600106 View Documents Alfa Aesar™
H6600106
5g Each for $351.61
 
Specifications

Chemical Identifiers

21947-97-7
387.494
AMTHPOXIVDHMLZ-KRWDZBQOSA-N
7015794
CC(C)(C)OC(=O)NC(CSC(C1=CC=CC=C1)C2=CC=CC=C2)C(=O)O
C21H25NO4S
MFCD00151869
boc-cys dpm-oh, boc-s-diphenylmethyl-l-cysteine, 2r-2-tert-butoxycarbonyl amino-3-diphenylmethyl sulfanyl propanoic acid, l-cysteine,n-1,1-dimethylethoxy carbonyl-s-diphenylmethyl
(2R)-3-benzhydrylsulfanyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid

Specifications

21947-97-7
MFCD00151869
AMTHPOXIVDHMLZ-KRWDZBQOSA-N
(2R)-3-benzhydrylsulfanyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
7015794
Solid
250mg
C21H25NO4S
boc-cys dpm-oh, boc-s-diphenylmethyl-l-cysteine, 2r-2-tert-butoxycarbonyl amino-3-diphenylmethyl sulfanyl propanoic acid, l-cysteine,n-1,1-dimethylethoxy carbonyl-s-diphenylmethyl
CC(C)(C)OC(=O)NC(CSC(C1=CC=CC=C1)C2=CC=CC=C2)C(=O)O
387.494
387.49
98%
S-Benzhydryl-N-Boc-L-cysteine