missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ (S)-(-)-Azetidine-2-carboxylic acid, 99%

Manufacturer:  Alfa Aesar™ L09668MF

 View more versions of this product

Catalog No. AAL09668MF



Specifications

Specifications

(S)-(-)-Azetidine-2-carboxylic acid
~217°C
2133-34-8
MFCD00005166
14,908
IADUEWIQBXOCDZ-VKHMYHEASA-N
(2S)-azetidine-2-carboxylic acid
16486
101.11
−123° (c=4 in H2O)
50mg
C4H7NO2
80678
l-azetidine-2-carboxylic acid, s-azetidine-2-carboxylic acid, s---2-azetidinecarboxylic acid, 2s-azetidine-2-carboxylic acid, azetidyl-2-carboxylic acid, l-azetidine 2-carboxylic acid, s-2-azetidinecarboxylic acid, 2-azetidinecarboxylic acid, s, s---azetidine-2-carboxylic acid, 2-azetidinecarboxylic acid, l
C1CNC1C(=O)O
101.105
CHEBI:6198
99%

Chemical Identifiers

2133-34-8
101.105
IADUEWIQBXOCDZ-VKHMYHEASA-N
16486
(2S)-azetidine-2-carboxylic acid
C4H7NO2
MFCD00005166
l-azetidine-2-carboxylic acid, s-azetidine-2-carboxylic acid, s---2-azetidinecarboxylic acid, 2s-azetidine-2-carboxylic acid, azetidyl-2-carboxylic acid, l-azetidine 2-carboxylic acid, s-2-azetidinecarboxylic acid, 2-azetidinecarboxylic acid, s, s---azetidine-2-carboxylic acid, 2-azetidinecarboxylic acid, l
CHEBI:6198
C1CNC1C(=O)O
Safety and Handling

Safety and Handling

RTECSNumber : CM4310500

Documents