Please login to your online account to display your discounted pricing

(S)-(-)-Atenolol, Tocris


β1-antagonist; Active isomer of atenolol

Quantity 10mg

Manufacturer: r & d systems  RDS039310

 View more versions of this product

Catalog No. 039310

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 10mg
Formula Weight 266.34
Chemical Name or Material (S)-(-)-Atenolol
Assay Percent Range >97%
Synonym (S)-(-)-4-[2-Hydroxy-3-[(1-methylethyl)amino]propoxy]benzeneacetamide
Molecular Formula C14H22N2O3
CAS 93379-54-5
Recommended Storage Store at Room Temperature

Specifications

Chemical Name or Material (S)-(-)-Atenolol
Assay Percent Range >97%
Molecular Formula C14H22N2O3
Formula Weight 266.34
Synonym (S)-(-)-4-[2-Hydroxy-3-[(1-methylethyl)amino]propoxy]benzeneacetamide
Accessories

Store at Room Temperature