Please login to your online account to display your discounted pricing

(S)-(-)-Atenolol, Tocris

β1-antagonist; Active isomer of atenolol

$133.98 - $133.98

Chemical Identifiers

Chemical Name or Material (S)-(-)-Atenolol
Assay Percent Range >97%
Molecular Formula C14H22N2O3
Formula Weight 266.34
Synonym (S)-(-)-4-[2-Hydroxy-3-[(1-methylethyl)amino]propoxy]benzeneacetamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

039310

 
r & d systems
RDS039310
10mg Each for $133.98
Description & Specifications

Specifications

Quantity 10mg
Formula Weight 266.34
Chemical Name or Material (S)-(-)-Atenolol
Assay Percent Range >97%
Synonym (S)-(-)-4-[2-Hydroxy-3-[(1-methylethyl)amino]propoxy]benzeneacetamide
Molecular Formula C14H22N2O3
CAS 93379-54-5
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature