Please login to your online account to display your discounted pricing

(S)-AMPA, Tocris

Selective AMPA agonist; Active isomer of (RS)-AMPA

$330.06 - $330.06

Chemical Identifiers

Chemical Name or Material (S)-AMPA
Assay Percent Range >99%
Molecular Formula C7H10N2O4
Formula Weight 186.17
Synonym (S)-α-Amino-3-hydroxy-5-methyl-4-isoxazolepropionic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

02-541-0

 
r & d systems
RDS025410
10mg Each for $330.06
Description & Specifications

Specifications

Quantity 10mg
Formula Weight 186.17
Chemical Name or Material (S)-AMPA
Assay Percent Range >99%
Molecular Formula C7H10N2O4
CAS 83643-88-3
Synonym (S)-α-Amino-3-hydroxy-5-methyl-4-isoxazolepropionic acid
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature