missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
(S)-(-)-α-Amino-γ-butyrolactone hydrobromide, 99%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 259940010
Specifications
(S)-(-)-α-Amino-γ-butyrolactone hydrobromide | |
Glass bottle | |
− 21.50 | |
236.0°C to 238.9°C | |
15295-77-9 | |
100.0 | |
C4H7NO2·BrH | |
Solubility in water: soluble. Other solubilities: soluble in ethanol,acetone | |
C1COC(=O)C1N.Br | |
182.02 | |
182.02 | |
99% |
Authentic | |
− 21.50 (20.00°C c=4,water) | |
Brown to Yellow | |
1g | |
98.5 | |
98.5 % min. (Argentometry) | |
2 3h-furanone, 3-aminodihydro-, hydrobromide, 3s, ksc924k2j, l---?-amino-?-butyrolactone hydrobromide, l---alpha-amino-gamma-butyrolactone hydrobromide, l-homoserine lactone hydrobromide, s---alpha-amino-gamma-butyrolactone hbr, s---alpha-amino-gamma-butyrolactone hydrobromide, s---alpha-amino-gamma-butyrolactonehydrobromide, s-2-amino-4-butyrolactone hydrobromide, s-3-aminodihydrofuran-2 3h-one hydrobromide | |
MKLNTBLOABOJFZ-DFWYDOINSA-N | |
(3S)-3-aminooxolan-2-one;hydrobromide | |
16213138 | |
Liquid |
Chemical Identifiers
15295-77-9 | |
182.02 | |
2 3h-furanone, 3-aminodihydro-, hydrobromide, 3s, ksc924k2j, l---?-amino-?-butyrolactone hydrobromide, l---alpha-amino-gamma-butyrolactone hydrobromide, l-homoserine lactone hydrobromide, s---alpha-amino-gamma-butyrolactone hbr, s---alpha-amino-gamma-butyrolactone hydrobromide, s---alpha-amino-gamma-butyrolactonehydrobromide, s-2-amino-4-butyrolactone hydrobromide, s-3-aminodihydrofuran-2 3h-one hydrobromide | |
(3S)-3-aminooxolan-2-one;hydrobromide |
C4H7NO2·BrH | |
MKLNTBLOABOJFZ-DFWYDOINSA-N | |
16213138 | |
C1COC(=O)C1N.Br |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF
GHS Signal Word: Warning