missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(S)-(-)-α-Amino-γ-butyrolactone hydrobromide, 99%, ACROS Organics™

$69.82 - $69.82

Chemical Identifiers

CAS 15295-77-9
Molecular Formula C4H7NO2·BrH
Molecular Weight (g/mol) 182.02
InChI Key MKLNTBLOABOJFZ-DFWYDOINSA-N
Synonym 2 3h-furanone, 3-aminodihydro-, hydrobromide, 3s, ksc924k2j, l---?-amino-?-butyrolactone hydrobromide, l---alpha-amino-gamma-butyrolactone hydrobromide, l-homoserine lactone hydrobromide, s---alpha-amino-gamma-butyrolactone hbr, s---alpha-amino-gamma-butyrolactone hydrobromide, s---alpha-amino-gamma-butyrolactonehydrobromide, s-2-amino-4-butyrolactone hydrobromide, s-3-aminodihydrofuran-2 3h-one hydrobromide
PubChem CID 16213138
IUPAC Name (3S)-3-aminooxolan-2-one;hydrobromide
SMILES C1COC(=O)C1N.Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC259940010
SDS View Documents
Thermo Fisher Scientific
259940010
1g Glass bottle
Each for $69.82
 
AC259940050
SDS View Documents
Thermo Fisher Scientific
AC259940050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

15295-77-9
182.02
2 3h-furanone, 3-aminodihydro-, hydrobromide, 3s, ksc924k2j, l---?-amino-?-butyrolactone hydrobromide, l---alpha-amino-gamma-butyrolactone hydrobromide, l-homoserine lactone hydrobromide, s---alpha-amino-gamma-butyrolactone hbr, s---alpha-amino-gamma-butyrolactone hydrobromide, s---alpha-amino-gamma-butyrolactonehydrobromide, s-2-amino-4-butyrolactone hydrobromide, s-3-aminodihydrofuran-2 3h-one hydrobromide
(3S)-3-aminooxolan-2-one;hydrobromide
C4H7NO2·BrH
MKLNTBLOABOJFZ-DFWYDOINSA-N
16213138
C1COC(=O)C1N.Br

Specifications

Authentic
− 21.50 (20.00°C c=4,water)
Brown to Yellow
1g
98.5
98.5 % min. (Argentometry)
2 3h-furanone, 3-aminodihydro-, hydrobromide, 3s, ksc924k2j, l---?-amino-?-butyrolactone hydrobromide, l---alpha-amino-gamma-butyrolactone hydrobromide, l-homoserine lactone hydrobromide, s---alpha-amino-gamma-butyrolactone hbr, s---alpha-amino-gamma-butyrolactone hydrobromide, s---alpha-amino-gamma-butyrolactonehydrobromide, s-2-amino-4-butyrolactone hydrobromide, s-3-aminodihydrofuran-2 3h-one hydrobromide
MKLNTBLOABOJFZ-DFWYDOINSA-N
(3S)-3-aminooxolan-2-one;hydrobromide
16213138
Liquid
(S)-(-)-α-Amino-γ-butyrolactone hydrobromide
Glass bottle
− 21.50
236.0°C to 238.9°C
15295-77-9
100.0
C4H7NO2·BrH
Solubility in water: soluble. Other solubilities: soluble in ethanol,acetone
C1COC(=O)C1N.Br
182.02
182.02
99%
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

GHS Signal Word: Warning

SDS