missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(S)-(-)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetic acid, 98%, ACROS Organics™

Manufacturer:  Acros Organics AC168230050

 View more versions of this product

Catalog No. AC168230050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Chiral derivatization reagent
Specifications

Specifications

(S)-(-)-α-Methoxy-α-(trifluoromethyl)phenylacetic acid
Authentic
1.4720 to 1.4750 (20°C, 589nm)
-69° to -73° (20°C, 589nm) (c=1.6, CH3OH)
5g
97.5
98%
C6H5C(OCH3)(CF3)CO2H
11,329
s---alpha-methoxy-alpha-trifluoromethyl phenylacetic acid, s-3,3,3-trifluoro-2-methoxy-2-phenylpropanoic acid, --mosher's acid, --mtpa, unii-172hcj1iqv, 172hcj1iqv, 2s-3,3,3-trifluoro-2-methoxy-2-phenylpropanoic acid, --alpha-methoxy-alpha-trifluoromethyl phenylacetic acid, s---mtpa, --a-methoxy-a-trifluoromethylphenylacetic acid
COC(C1=CC=CC=C1)(C(=O)O)C(F)(F)F
234.174
234.17
≥97.5%
1.3030g/mL
Glass bottle
- 71.00 (20.00°C c=1.6,CH3OH)
Colorless or White
17257-71-5
100.0
C10H9F3O3
MFCD00064200
15, 6365
JJYKJUXBWFATTE-VIFPVBQESA-N
(2S)-3,3,3-trifluoro-2-methoxy-2-phenylpropanoic acid
6992788
Viscous Liquid or Low Melting Solid

Chemical Identifiers

17257-71-5
234.174
JJYKJUXBWFATTE-VIFPVBQESA-N
6992788
COC(C1=CC=CC=C1)(C(=O)O)C(F)(F)F
C10H9F3O3
MFCD00064200
s---alpha-methoxy-alpha-trifluoromethyl phenylacetic acid, s-3,3,3-trifluoro-2-methoxy-2-phenylpropanoic acid, --mosher's acid, --mtpa, unii-172hcj1iqv, 172hcj1iqv, 2s-3,3,3-trifluoro-2-methoxy-2-phenylpropanoic acid, --alpha-methoxy-alpha-trifluoromethyl phenylacetic acid, s---mtpa, --a-methoxy-a-trifluoromethylphenylacetic acid
(2S)-3,3,3-trifluoro-2-methoxy-2-phenylpropanoic acid
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

Recommended Storage : Refrigerator +4°C

SDS Documents