missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(S)-(-)-alpha-Hydroxy-gamma-butyrolactone 97.0+%, TCI America™

Manufacturer:  Tci America H0951100MG

Catalog No. H0951100MG



Description

Description

Specifications

Specifications

(S)-(-)-alpha-Hydroxy-gamma-butyrolactone
100mg
C4H6O3
(S)-(-)-Dihydro-3-hydroxy-2(3H)-furanone
C1COC(=O)C1O
102.089
102.09
≥97.0% (GC)
Yellow
52079-23-9
MFCD00211245
FWIBCWKHNZBDLS-VKHMYHEASA-N
(3S)-3-hydroxyoxolan-2-one
357853
Clear Liquid at 20°C

Chemical Identifiers

52079-23-9
102.089
FWIBCWKHNZBDLS-VKHMYHEASA-N
357853
C1COC(=O)C1O
C4H6O3
MFCD00211245
(S)-(-)-Dihydro-3-hydroxy-2(3H)-furanone
(3S)-3-hydroxyoxolan-2-one
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents