Please login to your online account to display your discounted pricing

Alfa Aesar™ (S)-alpha-Amino-3-hydroxy-5-methyl-4-isoxazolepropionic acid

Active enantiomer of AMPA that induces apoptosis in cortical neurons

Chemical Identifiers

Chemical Name or Material (S)-alpha-Amino-3-hydroxy-5-methyl-4-isoxazolepropionic acid
CAS 83643-88-3
Formula Weight 186.16
MDL Number MFCD00672630
Synonym (S)-2-Amino-3-(3-hydroxy-5-methylisoxazol-4-yl)propanoic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ65847LB0

 
alfa aesar™
J65847LB0
5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 186.16
CAS 83643-88-3
Chemical Name or Material (S)-alpha-Amino-3-hydroxy-5-methyl-4-isoxazolepropionic acid
Physical Form Powder
Synonym (S)-2-Amino-3-(3-hydroxy-5-methylisoxazol-4-yl)propanoic acid
Quantity 5mg
MDL Number MFCD00672630
Color White