Please login to your online account to display your discounted pricing

Alfa Aesar™ (S)-(-)-alpha-(4-Chlorophenyl)benzylamine L-(+)-tartrate salt, 97%

Chemical Identifiers

Chemical Name or Material (S)-(-)-alpha-(4-Chlorophenyl)benzylamine L-(+)-tartrate salt
CAS 163837-57-8
Formula Weight 367.78
MDL Number MFCD08752515
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6330103

 
alfa aesar™
H6330103
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6330106

 
alfa aesar™
H6330106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH63301MD

 
alfa aesar™
H63301MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 367.78
Storage Note 1 2° to 8°C
Assay 97%
Chemical Name or Material (S)-(-)-alpha-(4-Chlorophenyl)benzylamine L-(+)-tartrate salt
MDL Number MFCD08752515
Quantity 1g
CAS 163837-57-8
Physical Form Solid
Safety and Handling

2° to 8°C