Please login to your online account to display your discounted pricing

Alfa Aesar™ (S)-alpha-(4-Chlorophenyl)benzylamine, 98%

Chemical Identifiers

Chemical Name or Material (S)-alpha-(4-Chlorophenyl)benzylamine
CAS 163837-32-9
Formula Weight 217.69g/mol
Synonym (S)-(4-Chlorophenyl)(phenyl)methylamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6148203

 
alfa aesar™
H61482-03
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH61482MD

 
alfa aesar™
H61482-MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 1g
Chemical Name or Material (S)-alpha-(4-Chlorophenyl)benzylamine
CAS 163837-32-9
Physical Form Solid
Assay 98%
Formula Weight 217.69g/mol
Synonym (S)-(4-Chlorophenyl)(phenyl)methylamine