missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
(S)-(-)-alpha-Allylglycine, 98%, 98% ee, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 275412500
Specifications
(S)-(-)-α-Allylglycine | |
Authentic | |
− 37.00 | |
253.0°C to 255.0°C | |
98%, (98% e.e.) | |
97.5 | |
98% | |
H2C=CHCH2CH(NH2)CO2H | |
04, 2852 | |
Solubility in water: soluble. | |
C=CCC(C(=O)O)N | |
115.13 | |
115.13 | |
98%,98% ee |
97.5% min. | |
Glass bottle | |
White | |
250mg | |
16338-48-0 | |
100.0 | |
C5H9NO2 | |
MFCD00002627 | |
2s-2-aminopent-4-enoic acid, 3-vinyl-l-alanine, h-gly ally-oh, l-2-amino-4-pentenoic acid, l-allylglycine, s---2-amino-4-pentenoic acid, s-2-amino-4-pentenoic acid, s-2-aminopent-4-enoic acid, s-allylglycine | |
WNNNWFKQCKFSDK-BYPYZUCNSA-N | |
(2S)-2-aminopent-4-enoic acid | |
167529 | |
Crystalline Powder |
Chemical Identifiers
16338-48-0 | |
115.13 | |
WNNNWFKQCKFSDK-BYPYZUCNSA-N | |
167529 | |
C=CCC(C(=O)O)N |
C5H9NO2 | |
MFCD00002627 | |
2s-2-aminopent-4-enoic acid, 3-vinyl-l-alanine, h-gly ally-oh, l-2-amino-4-pentenoic acid, l-allylglycine, s---2-amino-4-pentenoic acid, s-2-amino-4-pentenoic acid, s-2-aminopent-4-enoic acid, s-allylglycine | |
(2S)-2-aminopent-4-enoic acid |
Safety and Handling
GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and
GHS Signal Word: Warning
TSCA : TSCA